Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-3-hydroxyglutarate 95%

Supplier: AMBEED, INC
Warning

A705803-100G A705803-25G A705803-500G A705803-5G
77006-000EA 50 USD InStock
77006-000 77006-002 77006-004 77006-006
Diethyl-3-hydroxyglutarate 95%
Diethyl-3-hydroxyglutarate
Precaución: for Research Use Only


Formula: C₉H₁₆O₅
MDL Number: MFCD00009202
CAS Number: 32328-03-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty