Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazole-5-amine 98%

Supplier: AMBEED, INC
Danger

A707738-10G A707738-25G A707738-5G
76992-732EA 17 USD InStock
76992-732 76992-734 76992-736
1H-Pyrazole-5-amine 98%
1H-Pyrazole-5-amine
Precaución: for Research Use Only


Formula: C₃H₅N₃
MW: 83.09 g/mol
MDL Number: MFCD00005236
CAS Number: 916420-28-5
UN: 3263
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty