Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluorotriphenylsilane 97%

Supplier: AMBEED, INC
Danger

A707937-1G A707937-250MG A707937-5G
77035-808EA 18 USD InStock
77035-808 77035-810 77035-812
Fluorotriphenylsilane 97%
Fluorotriphenylsilane
Precaución: for Research Use Only


Formula: C₁₈H₁₅FSi
MW: 278.4
Melting Pt: 63
Density: 1.212
MDL Number: MFCD00017899
CAS Number: 379-50-0
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty