Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,2S)-Cyclopropane-1,2-dicarboxylic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A708205-250MG A708205-1G
76992-738EA 132 USD InStock
76992-738 77684-622
(1S,2S)-Cyclopropane-1,2-dicarboxylic acid 95%, Ambeed.Inc
(1S,2S)-Cyclopropane-1,2-dicarboxylic acid
Precaución: For research use only.


Formula: C₅H₆O₄
MW: 130.09 g/mol
MDL Number: MFCD06227617
CAS Number: 14590-54-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty