Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-fluoro-3-nitrobenzoate 95%

Supplier: AMBEED, INC
Warning

A712475-100MG A712475-1G A712475-250MG A712475-5G
76992-770EA 8 USD InStock
76992-770 76992-772 76992-774 76992-776
tert-Butyl 4-fluoro-3-nitrobenzoate 95%
tert-Butyl 4-fluoro-3-nitrobenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₂FNO₄
MDL Number: MFCD16620871
CAS Number: 579514-75-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty