Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-8-(trifluoromethoxy)quinoline 98%

Supplier: AMBEED, INC
Warning

A713015-100MG A713015-1G A713015-250MG A713015-5G
76992-784EA 85 USD InStock
76992-784 76992-786 76992-788 76992-790
5-Bromo-8-(trifluoromethoxy)quinoline 98%
5-Bromo-8-(trifluoromethoxy)quinoline
Precaución: for Research Use Only


Formula: C₁₀H₅BrF₃NO
MDL Number: MFCD11504881
CAS Number: 1065074-23-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty