Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aristolochic acid A 99%

Supplier: AMBEED, INC
Danger

A714436-100MG A714436-1G A714436-1MG A714436-250MG A714436-25MG A714436-50MG
77035-846EA 108 USD InStock
77035-846 77035-848 77035-850 77035-852 77035-854 77035-856
Aristolochic acid A 99%
Aristolochic acid A
Precaución: for Research Use Only


Formula: C₁₇H₁₁NO₇
MDL Number: MFCD00004996
CAS Number: 313-67-7
UN: 1544
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty