Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Pyridyloxyacetic acid 98%

Supplier: AMBEED, INC
Warning

A717295-100MG A717295-1G A717295-250MG A717295-5G
77006-076EA 22 USD InStock
77006-076 77006-078 77006-080 77006-082
3-Pyridyloxyacetic acid 98%
3-Pyridyloxyacetic acid
Precaución: for Research Use Only


Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD04039772
CAS Number: 86649-57-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty