Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-amino-6-bromopicolinate 95%

Supplier: AMBEED, INC
Danger

A725126-100MG A725126-1G A725126-250MG A725126-5G
76992-878EA 13 USD InStock
76992-878 76992-880 76992-882 76992-884
Ethyl 3-amino-6-bromopicolinate 95%
Ethyl 3-amino-6-bromopicolinate
Precaución: for Research Use Only


Formula: C₈H₉BrN₂O₂
MDL Number: MFCD11656227
CAS Number: 28033-08-1
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty