Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(p-Tolyl)-1-naphthylamine 95+%

Supplier: AMBEED, INC
Warning

A727206-1G A727206-250MG A727206-5G
77035-944EA 44 USD InStock
77035-944 77035-946 77035-948
N-(p-Tolyl)-1-naphthylamine 95+%
N-(p-Tolyl)-1-naphthylamine
Precaución: for Research Use Only


Formula: C₁₇H₁₅N
MDL Number: MFCD00035709
CAS Number: 634-43-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty