Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-4-(trifluoromethyl)picolinonitrile 95%

Supplier: AMBEED, INC
Warning

A727533-100MG A727533-1G
77006-162EA 57 USD InStock
77006-162 77006-164
5-Chloro-4-(trifluoromethyl)picolinonitrile 95%
5-Chloro-4-(trifluoromethyl)picolinonitrile
Precaución: for Research Use Only


Formula: C₇H₂ClF₃N₂
MDL Number: MFCD18259806
CAS Number: 1156542-28-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty