Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-(trifluoromethyl)benzyl alcohol 95%

Supplier: AMBEED, INC
Warning

A736865-1G A736865-25G A736865-5G
77006-180EA 45 USD InStock
77006-180 77006-182 77006-184
2-Chloro-3-(trifluoromethyl)benzyl alcohol 95%
2-Chloro-3-(trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
MDL Number: MFCD01631537
CAS Number: 261763-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty