Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-1-methylazetidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A746548-10G A746548-1G A746548-25G A746548-5G A746548-250MG
76993-006EA 171 USD InStock
76993-006 76993-008 76993-010 76993-012 77614-294
3-Hydroxy-1-methylazetidine hydrochloride 98%
3-Hydroxy-1-methylazetidine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₉NO·ClH
MW: 123.581 g/mol
MDL Number: MFCD16605978
CAS Number: 26687-49-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty