Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Pivaloyloxy)methyl (2S,5R,6R)-6-((azepan-1-ylmethylene)amino)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A747031-10MG A747031-1MG A747031-25MG A747031-50MG
76993-014EA 58 USD InStock
76993-014 76993-016 76993-018 76993-020
(Pivaloyloxy)methyl (2S,5R,6R)-6-((azepan-1-ylmethylene)amino)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrochloride 98%
(Pivaloyloxy)methyl (2S,5R,6R)-6-((azepan-1-ylmethylene)amino)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₂₁H₃₄ClN₃O₅S
MDL Number: MFCD01682071
CAS Number: 32887-03-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty