Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Diformylphenylboronic acid 98%

Supplier: AMBEED, INC
Danger

A749208-100MG A749208-1G A749208-250MG A749208-5G
77019-012EA 20 USD InStock
77019-012 77019-014 77019-016 77019-018
3,5-Diformylphenylboronic acid 98%
3,5-Diformylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₇BO₄
MW: 177.9498 g/mol
MDL Number: MFCD03701570
CAS Number: 480424-62-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty