Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-cyclopentyl-2-hydroxy-2-phenylacetate 98%

Supplier: AMBEED, INC
Warning

A749209-10G A749209-1G A749209-5G
77006-240EA 48 USD InStock
77006-240 77006-242 77006-244
Methyl-2-cyclopentyl-2-hydroxy-2-phenylacetate 98%
Methyl-2-cyclopentyl-2-hydroxy-2-phenylacetate
Precaución: for Research Use Only


Formula: C₁₄H₁₈O₃
MDL Number: MFCD00019295
CAS Number: 19833-96-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty