Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Trifluormethyl)nicotinonitrile 99%

Supplier: AMBEED, INC
Danger

A752152-10G A752152-1G A752152-25G A752152-5G
77006-258EA 45 USD InStock
77006-258 77006-260 77006-262 77006-264
6-(Trifluormethyl)nicotinonitrile 99%
6-(Trifluormethyl)nicotinonitrile
Precaución: for Research Use Only


Formula: C₇H₃F₃N₂
MDL Number: MFCD03001347
CAS Number: 216431-85-5
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty