Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-bromo-4-methylbenzoate 98%

Supplier: AMBEED, INC
Warning

A752867-25G A752867-5G
76993-032EA 10 USD InStock
76993-032 76993-034
Ethyl 3-bromo-4-methylbenzoate 98%
Ethyl 3-bromo-4-methylbenzoate


Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD11042852
CAS Number: 147962-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty