Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,4-difluorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A754066-100MG A754066-1G A754066-250MG A754066-5G
76993-036EA 9 USD InStock
76993-036 76993-038 76993-040 76993-042
3-Bromo-2,4-difluorobenzoic acid 97%
3-Bromo-2,4-difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃BrF₂O₂
MDL Number: MFCD09261257
CAS Number: 651026-98-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty