Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzofuran-7-ylboronic acid 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A754594-1G A754594-5G
77019-056EA 51 USD InStock
77019-056 77684-688
Benzofuran-7-ylboronic acid 98%, Ambeed.Inc
Benzofuran-7-ylboronic acid
Precaución: For research use only.


Formula: C₈H₇BO₃
MDL Number: MFCD20488081
CAS Number: 1432610-21-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty