Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzofuran 96%

Supplier: AMBEED, INC
Warning

A758861-100MG A758861-250MG A758861-5G
77036-158EA 28 USD InStock
77036-158 77036-160 77670-044
4-Bromobenzofuran 96%
4-Bromobenzofuran
Precaución: for Research Use Only


Formula: C₈H₅BrO
MDL Number: MFCD09056755
CAS Number: 128868-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty