Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-4-((tert-Butoxycarbonyl)amino)cyclopent-2-enecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A759992-100MG A759992-250MG
77019-118EA 6 USD InStock
77019-118 77019-120
cis-4-((tert-Butoxycarbonyl)amino)cyclopent-2-enecarboxylic acid 95%
cis-4-((tert-Butoxycarbonyl)amino)cyclopent-2-enecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₇NO₄
MW: 227.25 g/mol
MDL Number: MFCD00211287
CAS Number: 108999-93-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty