Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-methylthieno[2,3-d]pyrimidine 97%

Supplier: AMBEED, INC
Warning

A765125-100MG A765125-1G A765125-250MG
77036-200EA 33 USD InStock
77036-200 77036-202 77036-204
2,4-Dichloro-5-methylthieno[2,3-d]pyrimidine 97%
2,4-Dichloro-5-methylthieno[2,3-d]pyrimidine
Precaución: for Research Use Only


Formula: C₇H₄Cl₂N₂S
MDL Number: MFCD09965519
CAS Number: 56844-38-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty