Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridazine-3-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A767994-10G A767994-1G A767994-250MG A767994-5G
77036-228EA 317 USD InStock
77036-228 77036-230 77036-232 77036-234
Pyridazine-3-carbonitrile 95%
Pyridazine-3-carbonitrile
Precaución: for Research Use Only


Formula: C₅H₃N₃
MDL Number: MFCD09881239
CAS Number: 53896-49-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty