Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A769184-1G A769184-250MG A769184-25G A769184-5G
77036-250EA 40 USD InStock
77036-250 77036-252 77036-254 77036-256
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 97%
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₇H₄ClN₃O
MDL Number: MFCD12406099
CAS Number: 908287-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty