Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-formylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A771195-100MG A771195-1G A771195-5G
76993-140EA 8 USD InStock
76993-140 76993-142 76993-144
3-Fluoro-4-formylbenzoic acid 98%
3-Fluoro-4-formylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₅FO₃
MW: 168.122 g/mol
MDL Number: MFCD20483546
CAS Number: 193290-80-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty