Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminotetrahydro-2H-thiopyran 98%

Supplier: AMBEED, INC
Warning

A775838-100MG A775838-1G A775838-250MG A775838-5G
77019-196EA 10 USD InStock
77019-196 77019-198 77019-200 77019-202
4-Aminotetrahydro-2H-thiopyran 98%
4-Aminotetrahydro-2H-thiopyran
Precaución: for Research Use Only


Formula: C₅H₁₁NS
MW: 117.213 g/mol
MDL Number: MFCD07371524
CAS Number: 21926-00-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty