Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-[(tert-butoxycarbonyl)amino]nicotinate 96%

Supplier: AMBEED, INC
Warning

A778141-10G A778141-1G A778141-250MG A778141-5G
77006-392EA 155 USD InStock
77006-392 77006-394 77006-396 77006-398
Methyl-6-[(tert-butoxycarbonyl)amino]nicotinate 96%
Methyl-6-[(tert-butoxycarbonyl)amino]nicotinate
Precaución: for Research Use Only


Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
MDL Number: MFCD05663797
CAS Number: 144186-11-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty