Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1-Methyl-1H-imidazol-4-yl)acetonitrile 95%

Supplier: AMBEED, INC
Warning

A781947-100MG A781947-1G A781947-250MG
77019-232EA 19 USD InStock
77019-232 77019-234 77019-236
2-(1-Methyl-1H-imidazol-4-yl)acetonitrile 95%
2-(1-Methyl-1H-imidazol-4-yl)acetonitrile
Precaución: for Research Use Only


Formula: C₆H₇N₃
MW: 121.13 g/mol
MDL Number: MFCD18808589
CAS Number: 41065-00-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty