Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-bromocrotonate 80%

Supplier: AMBEED, INC
Danger

A787584-10G A787584-1G A787584-25G A787584-5G
77006-470EA 132 USD InStock
77006-470 77006-472 77006-474 77006-476 77006-478
Methyl-4-bromocrotonate 80%
Methyl-4-bromocrotonate
Precaución: for Research Use Only


Formula: C₅H₇BrO₂
MDL Number: MFCD00000246
CAS Number: 1117-71-1
UN: 1760
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty