Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-3-hydroxybenzenesulfonate 95%

Supplier: AMBEED, INC
Warning

A797018-1G A797018-250MG A797018-5G
77006-546EA 30 USD InStock
77006-546 77006-548 77006-550
Sodium-3-hydroxybenzenesulfonate 95%
Sodium-3-hydroxybenzenesulfonate
Precaución: for Research Use Only


Formula: C₆H₅NaO₄S
MW: 196.16 g/mol
MDL Number: MFCD00464504
CAS Number: 14278-60-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty