Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-aminocyclopentanecarboxylate hydrochloride 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A799019-100MG A799019-1G A799019-250MG A799019-5G
77019-390EA 15 USD InStock
77019-390 77019-392 77019-394 77684-741
Methyl 3-aminocyclopentanecarboxylate hydrochloride 98%, Ambeed.Inc
Methyl 3-aminocyclopentanecarboxylate hydrochloride
Precaución: For research use only.


Formula: C₇H₁₄ClNO₂
MDL Number: MFCD22423158
CAS Number: 1398534-59-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty