Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[4-(Trifluoromethyl)phenyl]propylamine 97%

Supplier: AMBEED, INC
Danger

A801149-100MG A801149-1G A801149-250MG
77036-402EA 94 USD InStock
77036-402 77036-404 77036-406
1-[4-(Trifluoromethyl)phenyl]propylamine 97%
1-[4-(Trifluoromethyl)phenyl]propylamine
Precaución: for Research Use Only


Formula: C₁₀H₁₂F₃N
MDL Number: MFCD07784290
CAS Number: 885276-54-0
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty