Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Aminomethyl)-5-methylpyrazine 98%

Supplier: AMBEED, INC
Danger

A803027-100MG A803027-5G
77006-600EA 16 USD InStock
77006-600 77006-602
2-(Aminomethyl)-5-methylpyrazine 98%
2-(Aminomethyl)-5-methylpyrazine
Precaución: for Research Use Only


Formula: C₆H₉N₃
MDL Number: MFCD01314542
CAS Number: 132664-85-8
UN: 2922
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty