Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Benzyloxy-4-chloro-6-methoxyquinoline 98%

Supplier: AMBEED, INC
Warning

A803779-250MG A803779-25G A803779-5G
77036-426EA 10 USD InStock
77036-426 77036-428 77036-430
7-Benzyloxy-4-chloro-6-methoxyquinoline 98%
7-Benzyloxy-4-chloro-6-methoxyquinoline
Precaución: for Research Use Only


Formula: C₁₇H₁₄ClNO₂
MDL Number: MFCD12910463
CAS Number: 286371-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty