Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-phenylpyridine 95%

Supplier: AMBEED, INC
Danger

A803996-1G A803996-100MG A803996-250MG
77036-432EA 175 USD InStock
77036-432 77670-136 77670-138
4-Bromo-2-phenylpyridine 95%
4-Bromo-2-phenylpyridine
Precaución: for Research Use Only


Formula: C₁₁H₈BrN
MDL Number: MFCD11656282
CAS Number: 98420-98-5
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty