Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-4-fluoroaniline 98%

Supplier: AMBEED, INC
Danger

A805369-100MG A805369-1G A805369-250MG A805369-5G
76993-330EA 12 USD InStock
76993-330 76993-332 76993-334 76993-336
3,5-Dimethyl-4-fluoroaniline 98%
3,5-Dimethyl-4-fluoroaniline
Precaución: for Research Use Only


Formula: C₈H₁₀FN
MDL Number: MFCD09832247
CAS Number: 1840-27-3
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty