Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-hydroxy-3,5-diiodobenzoate 95%

Supplier: AMBEED, INC
Warning

A806568-1G A806568-250MG A806568-5G
77019-442EA 27 USD InStock
77019-442 77019-444 77019-446
Ethyl 4-hydroxy-3,5-diiodobenzoate 95%
Ethyl 4-hydroxy-3,5-diiodobenzoate
Precaución: for Research Use Only


Formula: C₉H₈I₂O₃
MW: 417.96 g/mol
MDL Number: MFCD00016423
CAS Number: 54073-94-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty