Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Octyloxy)acetic acid ≥98%

Supplier: AMBEED, INC
Danger

A808788-100MG A808788-1G A808788-250MG
76993-358EA 10 USD InStock
76993-358 76993-360 76993-362
2-(Octyloxy)acetic acid ≥98%
2-(Octyloxy)acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₂₀O₃
MDL Number: MFCD11196116
CAS Number: 63632-58-6
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty