Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-isopropoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A809194-100MG A809194-1G A809194-250MG A809194-5G
77019-448EA 19 USD InStock
77019-448 77019-450 77019-452 77019-454
2-Fluoro-3-isopropoxyphenylboronic acid 98%
2-Fluoro-3-isopropoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₂BFO₃
MDL Number: MFCD11504844
CAS Number: 855230-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty