Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-sulfamoylbenzoate 97%

Supplier: AMBEED, INC
Warning

A812216-100MG A812216-250MG
77036-486EA 113 USD InStock
77036-486 77036-488
Methyl-4-sulfamoylbenzoate 97%
Methyl-4-sulfamoylbenzoate
Precaución: for Research Use Only


Formula: C₈H₉NO₄S
MDL Number: MFCD03768500
CAS Number: 22808-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty