Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibromonaphthalene 98%

Supplier: AMBEED, INC
Warning

A814100-100MG A814100-1G A814100-250MG A814100-5G
77019-474EA 6 USD InStock
77019-474 77019-476 77019-478 77019-480
1,3-Dibromonaphthalene 98%
1,3-Dibromonaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₆Br₂
MDL Number: MFCD18411509
CAS Number: 52358-73-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty