Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Di-Boc-1,4-butanediamine 95%

Supplier: AMBEED, INC
Warning

A816495-100MG A816495-1G A816495-250MG
77019-500EA 6 USD InStock
77019-500 77019-502 77019-504
N,N'-Di-Boc-1,4-butanediamine 95%
N,N'-Di-Boc-1,4-butanediamine
Precaución: for Research Use Only


Formula: C₁₄H₂₈N₂O₄
MDL Number: MFCD21364426
CAS Number: 33545-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty