Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2-(4-Benzylpiperazin-1-yl)phenyl]methanol 98%

Supplier: AMBEED, INC
Warning

A816529-10G A816529-1G A816529-250MG A816529-5G
77006-670EA 177 USD InStock
77006-670 77006-672 77006-674 77006-676
[2-(4-Benzylpiperazin-1-yl)phenyl]methanol 98%
[2-(4-Benzylpiperazin-1-yl)phenyl]methanol
Precaución: for Research Use Only


Formula: C₁₈H₂₂N₂O
MW: 282.3801 g/mol
MDL Number: MFCD01764934
CAS Number: 261178-24-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty