Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-1H-pyrazole 98%

Supplier: AMBEED, INC
Warning

A816729-100MG A816729-1G A816729-250MG
77036-518EA 79 USD InStock
77036-518 77036-520 77036-522
3-Fluoro-1H-pyrazole 98%
3-Fluoro-1H-pyrazole
Precaución: for Research Use Only


Formula: C₃H₃FN₂
MDL Number: MFCD18205877
CAS Number: 14521-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty