Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Amino-1,3-benzoxazol-5-yl)boronic acid hydrochloride 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A817040-250MG A817040-1G
76993-408EA 53 USD InStock
76993-408 77684-769
(2-Amino-1,3-benzoxazol-5-yl)boronic acid hydrochloride 95%, Ambeed.Inc
(2-Amino-1,3-benzoxazol-5-yl)boronic acid hydrochloride
Precaución: For research use only.


Formula: C₇H₈BClN₂O₃
MDL Number: MFCD26405707
CAS Number: 1404480-15-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty