Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Morpholinophenyl)methanol 98%

Supplier: AMBEED, INC
Warning

A818915-100MG A818915-1G A818915-250MG A818915-5G
77019-512EA 13 USD InStock
77019-512 77019-514 77019-516 77019-518
(3-Morpholinophenyl)methanol 98%
(3-Morpholinophenyl)methanol
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₂
MDL Number: MFCD07772812
CAS Number: 145127-38-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty