Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-3-methyl-1H-pyrazol-4-ylamine dihydrochloride 95+%

Supplier: AMBEED, INC
Warning

A821891-1G A821891-250MG A821891-5G
77006-692EA 39 USD InStock
77006-692 77006-694 77006-696
1-Ethyl-3-methyl-1H-pyrazol-4-ylamine dihydrochloride 95+%
1-Ethyl-3-methyl-1H-pyrazol-4-ylamine dihydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₃Cl₂N₃
MDL Number: MFCD08456818
CAS Number: 1185293-13-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty