Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,1':3',1''-Terphenyl]-2'-amine 95%

Supplier: AMBEED, INC
Danger

A830710-10G A830710-250MG A830710-5G A830710-1G
77019-576EA 181 USD InStock
77019-576 77019-578 77019-580 77573-648
[1,1':3',1''-Terphenyl]-2'-amine 95%
[1,1':3',1''-Terphenyl]-2'-amine
Precaución: for Research Use Only


Formula: C₁₈H₁₅N
MDL Number: MFCD09264304
CAS Number: 87666-57-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty