Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-N-(4-aminophenyl)benzenesulfonamide 95%

Supplier: AMBEED, INC
Danger

A831074-1G A831074-25G A831074-5G
77006-754EA 7 USD InStock
77006-754 77006-756 77006-758
4-Amino-N-(4-aminophenyl)benzenesulfonamide 95%
4-Amino-N-(4-aminophenyl)benzenesulfonamide
Precaución: for Research Use Only


Formula: C₁₂H₁₃N₃O₂S
MDL Number: MFCD00082174
CAS Number: 16803-97-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty